Potassium chloride 1 mol/l (1 N) in aqueous solution
Catalog # REGEKCL2105
Supplier: REAGECON
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360