wodorotlenek glinu, czysty
: CHEMPUR
:21645-51-2
:
:Al(OH)₃
:MFCD00003420
:12,00355
:300 °C
:2,42 g/cm³ (20 °C)
:78 g/mol
:244-492-7
:GHS07
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Woda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Chusteczki do twarzy
: VWR Collection
VWR®, Rękawice nitrylowe, NITRILE
: VWR Collection
VWR® NITRILE LIGHT, Rękawice nitrylowe
: VWR Collection
VWR®, Pipety Pasteura, High Performance
: VWR Collection
Rękawice nitrylowe, Kimtech™ Purple Nitrile™, Kimberly-Clark
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection