Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 30 - 32%, pure

Ammonia solution 30 - 32%, pure

Corrosive
Irritant
Environment
Supplier:   CHEMPUR
Ammonia solution 30 - 32%, pure
Supplier:  CHEMPUR
Certificates
Ammonia solution 30 - 32%, pure

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.