Aluminium potassium sulphate dodecahydrate, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7784-24-9
:
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143
:474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
EINECS:233-141-3
Frequently Bought Together