Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Ammonia solution 20%, pure

Corrosive
Irritant
Environment
Supplier:   CHEMPUR
Ammonia solution 20%, pure
Supplier:  CHEMPUR
Certificates
Ammonia solution 20%, pure

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 38,10 zł
    38,10 zł
    1 * 500 mL
       38,10 zł

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 45,60 zł
    45,60 zł
    1 * 1 L
       45,60 zł

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 189 zł
    189 zł
    1 * 5 L
       189 zł