Manganese(II) sulphate, anhydrous, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7785-87-7
:
Formula:MnO₄S
MDL Number:MFCD00003464
:151 g/mol
Merck Index:12,05782
Melting Pt:700…720 °C
Density:3.25 g/cm³ (20 °C)
EINECS:232-089-9
UN:3077
ADR:9,III
:GHS08
:GHS09
Frequently Bought Together