Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 10%, pure

Ammonia solution 10%, pure

Corrosive
Irritant
Environment
Supplier:   CHEMPUR
Ammonia solution 10%, pure
Supplier:  CHEMPUR
Certificates
Ammonia solution 10%, pure

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,70 zł
    31,70 zł
    1 * 500 mL
       31,70 zł

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,80 zł
    41,80 zł
    1 * 1 L
       41,80 zł

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 168 zł
    168 zł
    1 * 5 L
       168 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.