Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Copper reagent, CuVer 1, Powder Pillows
Copper reagent, CuVer 1, Powder Pillows
Copper reagent, CuVer 1, Powder Pillows
Catalog # 705-0089
Supplier:
Hach
CAS Number:
Rozwiń obraz
Copper reagent, CuVer 1, Powder Pillows
Catalog # 705-0089
Supplier:
Hach
CAS Number:
Specifications
Chemical method:
Bicinchoninate
Pk:
100
Description:
Copper
Type:
PP
Range:
0,04 - 5,00 mg/l
Specifications
About this item
Product Details
Documentation
Product Details
Documentation