Benzo[a]acridine
Catalog # PROOBCR-157
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:229.28 g/mol
EINECS:205-929-7
CAS Number:225-11-6
Formula:C₁₇H₁₁N