1-Aza-15-crown-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecane)
Catalog # MOLEM23329413
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.28 g/mol
EINECS:266-523-3
CAS Number:66943-05-3
Formula:C₁₀H₂₁NO₄
MDL Number:MFCD00075465