Order Entry
Poland
ContactUsLinkComponent
6,9-Dichloro-2-methoxyacridine, Sigma-Aldrich®
6,9-Dichloro-2-methoxyacridine, Sigma-Aldrich®
Catalog # SIAL145688-25G
Supplier:  Merck
CAS Number:  
6,9-Dichloro-2-methoxyacridine, Sigma-Aldrich®
Catalog # SIAL145688-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :278.14 g/mol
EINECS:201-666-7
CAS Number:86-38-4
Formula:C₁₄H₉Cl₂NO
MDL Number:MFCD00005028