(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid, Sigma-Aldrich®
Catalog # SIAL474533-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:182.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:407-960-3
CAS Number:94050-90-5
Formula:C₉H₁₀O₄
MDL Number:MFCD00274088