Methanol, super gradient grade for HPLC
POCH621995156-2.5L
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
:
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:11 °C (closed cup)
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS02
:GHS06
:GHS08
:67-56-1
:CH₃OH
:MFCD00004595