
Sodium hydroxide 0.1 mol concentrated aqueous solution volumetric solution
Catalog # CHMP168109336.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548