L(+)-Ascorbic acid, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Jump to:
About this item
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
5 Options Available Below
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



