
3-Phenylpyridine ≥97.0% for synthesis
Catalog # 8.41759.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Assay (GC, area%):≥ 97.0 % (a/a)
- Density (d 20 °C/ 4 °C):1.090 - 1.094
- Identity (IR):passes test
Specification Test Results
About this item
:
:155.2 g/mol
Boiling Pt:273…274 °C (1013 hPa)
Melting Pt:76…77 °C
Density:1.08 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:213-762-6
:GHS07
CAS Number:1008-88-4
Formula:C₁₁H₉N
MDL Number:MFCD00006380