
3-Aminopirydyna
Catalog # MOLE20122882-500G
Supplier: Molekula
CAS Number:
Specifications
- Rodzaj opakowania:Glass bottle for solids
- Ilość:500 g
- Seq:3645
Specifications
About this item
:
:94.12 g/mol
Merck Index:13,00472
Boiling Pt:∼248 °C (1013 hPa)
Melting Pt:60…63 °C
Flash Pt:88 °C
Storage Temperature:Temperatura otoczenia
EINECS:207-322-2
UN:2671
ADR:6.1,II
:GHS06
CAS Number:462-08-8
Formula:C₅H₆N₂
MDL Number:MFCD00006400