Order Entry
Poland
ContactUsLinkComponent
Sterilin® Petri Dishes
Sterilin® Petri Dishes
Catalog # 391-2028
Supplier:  STERILIN
undefined
Sterilin® Petri Dishes
Catalog # 391-2028
Supplier:  STERILIN

Specifications

  • Ø:
    139 mm
  • Sterility:
    Non sterile
  • Version:
    140 mm, triple vent
  • Height:
    21,1 mm
  • Pk:
    80

Specifications

About this item

Ideal for applications where a large surface area and very flat base are required.

  • Easy grip ridges on the base to aid individual dish handling
  • Triple vented to aid gaseous exchange
  • Ideal for use in automatic plate pourers

Dimensions shown are for outer Ø base and outer height (mm).

Certifications: Produced and tested in accordance with EN ISO 24998