
Manganese(IV) telluride ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12032-89-2
:
Formula:MnTe₂
MDL Number:MFCD00064703
:310.14 g/mol
EINECS:234-783-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together