

Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-11-6
:
Formula:Cl₄Zr
MDL Number:MFCD00011306
:233.03 g/mol
Merck Index:14,10174
Boiling Pt:3577 °C (1013 hPa)
Melting Pt:331 °C (subl.)
Density:2.803
Storage Temperature:Ambient
EINECS:233-058-2
UN:2503
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together