

Zirconium tetrachloride, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-11-6
:
Formula:Cl₄Zr
MDL Number:MFCD00011306
:233.03 g/mol
Merck Index:12,10307
Boiling Pt:3577 °C (1013 hPa)
Melting Pt:437 °C (subl.)
Density:2,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-058-2
UN:2503
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White Powder
- Assay:≥97.5 % (ZrCl4)
- Aluminium (Al):≤100 ppm
- Chromium (Cr):≤100 ppm
- Hafnium (Hf):≤100 ppm
- Iron (Fe):≤1000 ppm
- Silicon (Si):≤150 ppm
- Titanium (Ti):≤100 ppm
Specification Test Results
Frequently Bought Together