Order Entry
Poland
ContactUsLinkComponent
Silver nitrate ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Corrosive
Oxidizing
Environment
Silver nitrate ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Certificates
undefined
Corrosive
Oxidizing
Environment
Silver nitrate ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.8 %
  • Acid. or alk. react. impurities:
    complying
  • Free acid:
    complying
  • Cadmium (Cd):
    Max. 0.0001 %
  • Copper (Cu):
    Max. 0.0002 %
  • Iron (Fe):
    Max. 0.0002 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.001 %
  • Zinc (Zn):
    Max. 0.0001 %
  • Substances not precipitated by HCl (as SO₄):
    Max. 0.01 %
  • Chloride (Cl):
    Max. 0.0001 %
  • Nitrite (NO₂):
    Max. 0.05 %
  • Sulfate (SO₄):
    Max. 0.002 %
  • Al, Pb, Bi and Cu ions:
    complying
  • Appearance of the solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together