Lanthanum(III) oxide, Sigma-Aldrich®
Catalog # SIAL203556-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:325.81 g/mol
Boiling Pt:4200 °C (1013 hPa)
Melting Pt:2315 °C
Density:6.51 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-200-5
CAS Number:1312-81-8
Formula:La₂O₃
MDL Number:MFCD00011071