Order Entry
Poland
ContactUsLinkComponent
Lanthanum(III) oxide, Supelco®
Lanthanum(III) oxide, Supelco®
Supplier:  Merck
Certificates
Lanthanum(III) oxide, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    2315 °C
  • Molar Mass:
    325.82 g/mol
  • Bulk Density:
    1300 kg/m3
  • Boiling Point:
    4200 °C
  • Density:
    6.51 g/cm3 (20 °C)
  • pH:
    9.0 (50 g/l, H2O, 20 °C) (slurry)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together