
Bismuth(III) chloride, anhydrous 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-60-2
:
Formula:BiCl₃
MDL Number:MFCD00003461
:315.34 g/mol
Merck Index:13,01261
Boiling Pt:447 °C (760 mmHg)
Melting Pt:230…232 °C
Density:4.75 g/cm³
Storage Temperature:Ambient
EINECS:232-123-2
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White to beige to grey Crystalline powder and/or chunks
- Titration Argentometric:≥98.0 %
- Water:≤3 % (K.F.)
Specification Test Results
Frequently Bought Together