


Antimony(V) chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-18-9
:
Formula:Cl₅Sb
MDL Number:MFCD00011213
:299.02 g/mol
Merck Index:14,695
Boiling Pt:79 °C (22 mmHg)
Melting Pt:2.8 °C
Density:2.336
Storage Temperature:Ambient
UN:1730
ADR:8,II
:ampouled under argon
EINECS:231-601-8
:GHS05
:GHS07
:GHS09
Related Information
Frequently Bought Together