Chloroacetamidine hydrochloride, Sigma-Aldrich®
Catalog # SIAL591475-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:128.99 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:233-675-7
UN:3261
CAS Number:10300-69-3
Formula:C₂H₆Cl₂N₂
MDL Number:MFCD00053013