2-Hydrazinobenzoic acid hydrochloride, Sigma-Aldrich®
Catalog # SIAL324302-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:188.61 g/mol
:M-Clarity™ quality level = MQ100
EINECS:257-869-6
CAS Number:52356-01-1
Formula:C₇H₉ClN₂O₂
MDL Number:MFCD00012931