Bis(cyclopentadienyl)titanium dichloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1271-19-8
:
Formula:C₁₀H₁₀Cl₂Ti
MDL Number:MFCD00003723
:248.97 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:215-035-9
UN:3261
Frequently Bought Together