1,3-Dibromo-5,5-dimethylhydantoin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-48-5
:
Formula:C₅H₆Br₂N₂O₂
MDL Number:MFCD00003189
:M-Clarity™ quality level = MQ200
:285.92 g/mol
EINECS:201-030-9
Frequently Bought Together