Order Entry
Poland
ContactUsLinkComponent
3-Methyl-1-phenyl-2-pyrazolin-5-one
3-Methyl-1-phenyl-2-pyrazolin-5-one
Catalog # MOLEM79333761
Supplier:  Molekula
CAS Number:  
3-Methyl-1-phenyl-2-pyrazolin-5-one
Catalog # MOLEM79333761
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :174.2 g/mol
EINECS:201-891-0
CAS Number:89-25-8
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138