Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
Poland
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
ContactUsLinkComponent
Home
(R)-(-)-β-Aminophenethyl alcohol
(R)-(-)-β-Aminophenethyl alcohol
(R)-(-)-β-Aminophenethyl alcohol
8.41290.0001
:
Merck
:
(R)-(-)-β-Aminophenethyl alcohol
8.41290.0001
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
137.18 g/mol
:
260-287-5
:
56613-80-0
:
C₈H₁₁NO
:
MFCD00008062