

Acetonitrile for HPLC
POCH102640150-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
:
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45.7 °C
:0.782 g/cm³ (20 °C)
:2 °C (closed cup)
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS02
:GHS07
:75-05-8
:H₃CCN
:MFCD00001878