Order Entry
Poland
ContactUsLinkComponent
Organic reference standard, Hexobarbital 1,000 µg/ml in methanol, Cerilliant
Organic reference standard, Hexobarbital 1,000 µg/ml in methanol, Cerilliant
  PROOCERH-013
 :  
Organic reference standard, Hexobarbital 1,000 µg/ml in methanol, Cerilliant
  PROOCERH-013
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    56-29-1
  • Description:
    Hexobarbital 1,000 µg/ml in methanol
  • Environmentally Preferable:
  • Formula:
    C₁₂H₁₆N₂O₃
  • Description:
    Hexobarbital