Order Entry
Poland
ContactUsLinkComponent
cis-2,3-Epoxybutane 97%
Irritant
Flammable
cis-2,3-Epoxybutane 97%
Certificates
undefined
Irritant
Flammable
cis-2,3-Epoxybutane 97%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3820 to 1.3840 (20°C, 589 nm)
  • Water:
    ≤2.5 %
  • GC:
    ≥96.0 % (corrected for H2O)

Specification Test Results

Related Information

Frequently Bought Together