



3-Chloropropene 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-05-1
:
Formula:C₃H₅Cl
MDL Number:MFCD00000984
:76.53 g/mol
Merck Index:13,00286
Boiling Pt:44…46 °C (760 mmHg)
Melting Pt:–136 °C
Density:0.939 g/cm³
Flash Pt:–29 °C
UN:1100
ADR:3,I
:Stabilised with propylene oxide
EINECS:203-457-6
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4135 to 1.4175 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:0.05 to 0.09 % Propylene oxide
Specification Test Results
Frequently Bought Together