
Sulphuric acid ≥96%, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # 6902.0500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay (H₂SO₄)(w/w):93 - 98 %
- Trace Impurities - Aluminum (Al):≤ 50 ppt
- Trace Impurities - Antimony (Sb):≤ 50 ppt
- Trace Impurities - Arsenic (As):≤ 500 ppt
- Trace Impurities - Barium (Ba):≤ 10 ppt
- Trace Impurities - Beryllium (Be):≤ 10 ppt
- Trace Impurities - Bismuth (Bi):≤ 10 ppt
- Trace Impurities - Cadmium (Cd):≤ 10 ppt
- Trace Impurities - Calcium (Ca):≤ 50 ppt
- Trace Impurities - Cerium (Ce):≤ 10 ppt
- Trace Impurities - Cesium (Cs):≤ 10 ppt
- Trace Impurities - Chromium (Cr):≤ 10 ppt
- Trace Impurities - Cobalt (Co):≤ 10 ppt
- Trace Impurities - Copper (Cu):≤ 10 ppt
- Trace Impurities - Dysprosium (Dy):≤ 10 ppt
- Trace Impurities - Erbium (Er):≤ 10 ppt
- Trace Impurities - Europium (Eu):≤ 10 ppt
- Trace Impurities - Gadolinium (Gd):≤ 10 ppt
- Trace Impurities - Gallium (Ga):≤ 10 ppt
- Trace Impurities - Germanium (Ge):≤ 100 ppt
- Trace Impurities - Hafnium (Hf):≤ 10 ppt
- Trace Impurities - Holmium (Ho):≤ 10 ppt
- Trace Impurities - Indium (In):≤ 10 ppt
- Trace Impurities - Iron (Fe):≤ 50 ppt
- Trace Impurities - Lanthanum (La):≤ 10 ppt
- Trace Impurities - Lead (Pb):≤ 10 ppt
- Trace Impurities - Lithium (Li):≤ 10 ppt
- Trace Impurities - Lutetium (Lu):≤ 10 ppt
- Trace Impurities - Magnesium (Mg):≤ 50 ppt
- Trace Impurities - Manganese (Mn):≤ 10 ppt
- Trace Impurities - Mercury (Hg):≤ 100 ppt
- Trace Impurities - Molybdenum (Mo):≤ 10 ppt
- Trace Impurities - Neodymium (Nd):≤ 10 ppt
- Trace Impurities - Nickel (Ni):≤ 50 ppt
- Trace Impurities - Niobium (Nb):≤ 10 ppt
- Trace Impurities - Potassium (K):≤ 50 ppt
- Trace Impurities - Praseodymium (Pr):≤ 10 ppt
- Trace Impurities - Rhodium (Rh):≤ 50 ppt
- Trace Impurities - Rubidium (Rb):≤ 10 ppt
- Trace Impurities - Samarium (Sm):≤ 10 ppt
- Trace Impurities - Scandium (Sc):≤ 10 ppt
- Trace Impurities - Selenium (Se):≤ 500 ppt
- Trace Impurities - Silver (Ag):≤ 50 ppt
- Trace Impurities - Sodium (Na):≤ 50 ppt
- Trace Impurities - Strontium (Sr):≤ 10 ppt
- Trace Impurities - Tellurium (Te):≤ 50 ppt
- Trace Impurities - Terbium (Tb):≤ 10 ppt
- Trace Impurities - Thallium (Tl):≤ 10 ppt
- Trace Impurities - Thorium (Th):≤ 10 ppt
- Trace Impurities - Thulium (Tm):≤ 10 ppt
- Trace Impurities - Tin (Sn):≤ 50 ppt
- Trace Impurities - Titanium (Ti):≤ 50 ppt
- Trace Impurities - Tungsten (W):≤ 10 ppt
- Trace Impurities - Uranium (U):≤ 10 ppt
- Trace Impurities - Vanadium (V):≤ 10 ppt
- Trace Impurities - Ytterbium (Yb):≤ 10 ppt
- Trace Impurities - Yttrium (Y):≤ 10 ppt
- Trace Impurities - Zinc (Zn):≤ 50 ppt
- Trace Impurities - Zirconium (Zr):≤ 10 ppt
Specification Test Results
About this item
:
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589