

Sodium metabisulphite, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0267.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:min. 97.0 %
- Assay (SO2):min. 58.5 %
- Chloride (Cl):max. 0.005 %
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble Matter:max. 0.005 %
- Thiosulfate (S2O3):max. 0.05 %
- Iron (Fe):max. 5
Specification Test Results
About this item
:
:190.11 g/mol
Merck Index:12,08784
Melting Pt:150 °C
Density:1.48 g/cm³ (20 °C)
EINECS:231-673-0
REACH:01-2119531326-45
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7681-57-4
Formula:Na₂S₂O₅
MDL Number:MFCD00167602