

Sodium peroxide ≥95%, Puriss. p.a. ACS, ISO, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1313-60-6
:
Formula:Na₂O₂
MDL Number:MFCD00003497
:77.98 g/mol
Merck Index:13,08728
Boiling Pt: Dec. (on heating)
Melting Pt:660 °C
Density:2.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-209-4
UN:1504
ADR:5.1,I
:GHS05
:GHS03
Specification Test Results
- Assay:Min. 95.0 %
- Aluminium (Al):Max. 0.001 %
- Iron (Fe):Max. 0.002 %
- Heavy metals (as Pb):Max. 0.002 %
- Chloride (Cl):Max. 0.002 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.001 %
- Total N:Max. 0.003 %
Specification Test Results
Frequently Bought Together