Sodium amide 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-92-5
:
Formula:H₂NNa
MDL Number:MFCD00011117
:39.01 g/mol
Merck Index:12,08717
Boiling Pt:400 °C
Melting Pt:210 °C
Density:1,39 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-971-0
UN:3131
ADR:4.3,I
:GHS05
:GHS02
:GHS09
Specification Test Results
- Appearance:White to greyish-white Powder or irregular granules or flakes
- Titration with HClO4:≥98.5 %
- Free Sodium:Passes test (A small amount dropped into water, does not catch fire)
- Peroxides:≤0.1 % (as NaNH2O2)
Specification Test Results
Frequently Bought Together