

Sodium azide
Catalog # MOLE31803515-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:65.01 g/mol
Merck Index:13,08653
Melting Pt:275 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:247-852-1
UN:1687
ADR:6.1,II
:GHS06
:GHS09
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536