Order Entry
Poland
ContactUsLinkComponent
 
Potassium iodide, Sigma-Aldrich®
Catalog # SIAL60422-250G
Supplier:  Merck
CAS Number:  
Potassium iodide, Sigma-Aldrich®
Catalog # SIAL60422-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:686 °C
Density:3.13 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405