Order Entry
Poland
ContactUsLinkComponent
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Acidity:
    Max. 0.05 meq/g
  • Alkalinity:
    Max. 0.02 meq/g
  • Heavy metals (as Pb):
    Max. 30 ppm
  • Insolubility in water:
    Max. 0.1 %
  • Cl (Chloride):
    Max. 0.05 %
  • SiF6 (Hexafluorosilicate):
    Max. 0.3 %
  • Fe (Iron):
    Max. 30 ppm

Specification Test Results

Frequently Bought Together