Potassium fluoride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7789-23-3
:
Formula:KF
MDL Number:MFCD00011398
:58.1 g/mol
Merck Index:13,07714
Boiling Pt:1505 °C (1013 hPa)
Melting Pt:860 °C
Density:2.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-151-5
UN:1812
ADR:6.1,III
:GHS06
Specification Test Results
- Assay:Min. 99.0 %
- Acidity:Max. 0.025 meq/g
- Alkalinity:Max. 0.01 meq/g
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 50 ppm
- SiF6 (Hexafluorosilicate):Max. 0.05 %
- SO4 (Sulphate):Max. 0.05 %
- Cu (Copper):Max. 25 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 25 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC26820.236 spec_VWRC26820.236 spec_VWRC26820.293 spec_VWRC26820.293
Product specification
Frequently Bought Together