Potassium fluoride, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7789-23-3
:
Formula:KF
MDL Number:MFCD00011398
:58.1 g/mol
Merck Index:13,07714
Boiling Pt:1505 °C (1013 hPa)
Melting Pt:860 °C
Density:2.48 g/cm³ (20 °C)
UN:1812
ADR:6.1,III
:M-Clarity™ quality level = MQ300
EINECS:232-151-5
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:923 g/l (20 °C)
- Melting Point:860 °C
- Molar Mass:58.1 g/mol
- Bulk Density:400 kg/m3
- Boiling Point:1500 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (885 °C)
- Density:2.48 g/cm3 (20 °C)
- pH:7 - 9 (50 g/l, H2O, 18 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together