Order Entry
Poland
ContactUsLinkComponent
Potassium fluoride ≥99.9% (metals basis)
Potassium fluoride ≥99.9% (metals basis)
Catalog # 42217.14
CAS Number:  
Potassium fluoride ≥99.9% (metals basis)
Catalog # 42217.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :58.1 g/mol
Merck Index:14,7632
Boiling Pt:1505 °C
Melting Pt:859 °C
Density:2.48
Storage Temperature:Ambient
EINECS:232-151-5
UN:1812
ADR:6.1,III
 :GHS06
CAS Number:7789-23-3
Formula:KF
MDL Number:MFCD00011398