Order Entry
Poland
ContactUsLinkComponent
Potassium dichromate ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Potassium dichromate ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Potassium dichromate ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 99.8 %
  • Insolubility in water:
    Max. 0.03 %
  • Loss on drying (130°C):
    Max. 0.05 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 200 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Na (Sodium):
    Max. 0.1 %
  • Pb (Lead):
    Max. 50 ppm
  • Statement on the use:
    Confirmed
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together