


Sodium perchlorate 98%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7601-89-0
:
Formula:ClNaO₄
MDL Number:MFCD00003480
:122.44 g/mol
Merck Index:13,08726
Melting Pt:482 °C
Density:2,4989 to 2.4999 g/cm³ (25 °C)
EINECS:231-511-9
UN:1502
ADR:5.1,II
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or crystals and/or chunks
- pH:6.0 to 8.0 (5% in water, 25°C)
- Titration after Ion exchange:97.5 to 102.5 %
- Insoluble matter in water:≤0.005 %
Specification Test Results
Related Information
Frequently Bought Together