Order Entry
Poland
ContactUsLinkComponent
Lithium perchlorate ≥95.0% ACS, Fluka™
Irritant
Oxidizing
Lithium perchlorate ≥95.0% ACS, Fluka™
Catalog # 205281-100G
CAS Number:  
Irritant
Oxidizing
Lithium perchlorate ≥95.0% ACS, Fluka™
Catalog # 205281-100G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 95.0 %
  • pH (5 %, 25 °C):
    6.0 - 7.5
  • Water insoluble matter:
    Max. 0.005 %
  • Iron (Fe):
    Max. 5 ppm
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.003 %
  • Sulfate (SO₄):
    Max. 0.001 %

Specification Test Results

About this item

 :
 :106.39 g/mol
Merck Index:13,05561
Boiling Pt:430 °C (1013 hPa)
Melting Pt:236 °C
Density:2.420 g/cm³
Storage Temperature:Ambient
EINECS:232-237-2
UN:1481
ADR:5.1,II
 :GHS07
 :GHS03
CAS Number:7791-03-9
Formula:ClLiO₄
MDL Number:MFCD00011079