Magnetic stirring bars, PTFE, tandem, BOLA
Tandem shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE. The centre bore structure is used for receiving bearing neck or a glass neck. In these stirrer bars reduction of running surface to a ring minimises friction and increases lifespan.
- Chemical resistant
- Extremely strong mixing can be achieved
- Ideal transmission of the magnetic force of the stirrer to the tandem magnetic stirring bar
Temperature resistance: −200 to +250 °C.
Frequently Bought Together